Bromthymolblau -Lösung R3 Reag. Ph. Eur.

Lieferant: VWR Chemicals

84118.180EA 78.4 EUR
84118.180
Bromthymolblau -Lösung R3 Reag. Ph. Eur.
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
MDL: MFCD00005872
CAS-Nummer: 76-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR