Aceton ≥99,9%, SPECTRONORM® für die Spektroskopie

Lieferant: VWR Chemicals
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

Filtriert über 0,2 µm-Filter. Unter Stickstoff abgefüllt.

84700.320EA 143 EUR
84700.320 84700.260
Aceton ≥99,9%, SPECTRONORM® für die Spektroskopie
Aceton

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥ 99.9 %
Solubility in water Passes test ACS
Substances reducing KMnO₄ (as O) Passes test ACS
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Colouration ≤ 10 APHA
Aldehydes (as HCHO) ≤ 10 ppm
Evaporation residue ≤ 5 ppm
2-Propanol ≤ 0.05 %
Methanol ≤ 0.05 %
Water ≤ 0.2 %
Absorbance (330 nm) ≤ 0.70
Absorbance (335 nm) ≤ 0.22
Absorbance (340 nm) ≤ 0.07
Absorbance (345 nm) ≤ 0.02
Absorbance (from 350 nm) ≤ 0.004
Transmittance (330 nm) ≥ 20 %
Transmittance (335 nm) ≥ 60 %
Transmittance (340 nm) ≥ 85 %
Transmittance (345 nm) ≥ 95 %
Transmittance (from 350 nm) ≥ 99 %
Fluorescence (as quinine) (365 nm) ≤ 1.0 ppb
Meets the requirements of "Specific use spectroscopy" according ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR