Chrom(III)chlorid Hexahydrat 98-101%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Achtung
|
84839.260EA
79.3
EUR
84839.260
Chrom(III)chlorid Hexahydrat 98-101%, GPR RECTAPUR®
Chrom(III)chlorid Hexahydrat
Formel:
CrCl₃·6H₂O Schmelzpunkt: 85 °C Dichte: 1,76 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149660 CAS-Nummer: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8,III Merck Index: 13,02242 |
Spezifikation Testergebnisse
Assay | 98 - 101 % |
Insolubility in water | Max. 100 ppm |
Substances not precipitated by NH4OH | Max. 0.2 % |
SO4 (Sulphate) | Max. 100 ppm |
Fe (Iron) | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...