Kupfer(II)sulfat, wasserfrei ≥99,0%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals
Achtung

Synonyme: Kupfersulfat

84845.230EA 64.8 EUR
84845.230 84845.290
Kupfer(II)sulfat, wasserfrei ≥99,0%, AnalaR NORMAPUR®
Kupfer(II)sulfat
Formel: CuSO₄
Schmelzpunkt: 200 °C
Dichte: 3,603 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010981
CAS-Nummer: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,02722

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) Min. 99.0 %
Identification Passes test
Insolubility in water Max. 100 ppm
Loss on drying (250°C) Max. 1 %
Substances not precipitated by H2S Max. 0.2 %
Total N (Nitrogen) Max. 20 ppm
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 50 ppm
Pb (Lead) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR