Natriumformiat ≥99%, AnalaR NORMAPUR®
Lieferant: VWR Chemicals
84850.260EA
64.4
EUR
84850.260
84850.460
Natriumformiat ≥99%, AnalaR NORMAPUR®
Natriumformiat
Formel:
HCO₂Na Schmelzpunkt: 257,31 °C Dichte: 1,919 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013101 CAS-Nummer: 141-53-7 EINECS: 205-488-0 REACH: 01-2119486468-21 Merck Index: 13,08694 |
Spezifikation Testergebnisse
Assay | Min. 99 % |
Heavy metals (as Pb) | Max. 5 ppm |
Insolubility in water | Max. 50 ppm |
Cl (Chloride) | Max. 10 ppm |
SO4 (Sulphate) | Max. 10 ppm |
Ca (Calcium) | Max. 50 ppm |
Fe (Iron) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...