Kupfer(II)acetat-Lösung USP Test Solution (TS)

Lieferant: VWR Chemicals

Synonyme: Essigsäure Kupfer(II)salz, Cupriaacetat, Kupferdiacetat

85341.180EA 26.5 EUR
85341.180
Kupfer(II)acetat-Lösung USP Test Solution (TS)
Kupfer(II)acetat

Ready to use Cupric acetate test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formel: CuCH₃CO₂
Molecular Weight: 181,64 g/mol
Schmelzpunkt: 115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008690
CAS-Nummer: 142-71-2
EINECS: 0
ADR: 9,III
Merck Index: 12,02690

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR