Resorcin USP Test Solution (TS)

Lieferant: VWR Chemicals

Synonyme: 1,3-Dihydroxybenzol, Resorcinol

85369.180EA 147 EUR
85369.180
Resorcin USP Test Solution (TS)
Resorcin

Ready to use test solutions (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formel: C₆H₆O₂
Molecular Weight: 110,11 g/mol
Schmelzpunkt: 109 bis 111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002269
CAS-Nummer: 108-46-3
EINECS: 0
ADR: 6.1,III
Merck Index: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR