tri-Natriumcitrat-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

854-200ML
854-200MLEA 118 EUR
854-200ML
tri-Natriumcitrat-Lösung, Sigma-Aldrich®
tri-Natriumcitrat

Citrate Solution for the preparation of a 60% citrate buffered acetone solution used as a fixative for blood smears. No dilution required. Add 30 ml citrate solution to 20 ml absolute acetone for preparation of fixative.

Formel: C₆H₅Na₃O₇
Lagertemperatur: Kühlschrank
MDL: MFCD00012462
CAS-Nummer: 68-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR