Salzsäure 5 mol/l (5 N) für die biopharmazeutische Produktion

Lieferant: VWR Chemicals
Achtung

Synonyme: Hydrochloric acid solution, Muriatic acid

85400.320EA 48 EUR
85400.320
Salzsäure 5 mol/l (5 N) für die biopharmazeutische Produktion
Salzsäure

This product is produced from hydrochloric acid 37 % Ph.Eur. and water Ph.Eur., USP, NF purified.


Residues of metal catalysts or reagents according to EMEA/CHMP/SWP/4446/2000 are not likely to be present.

Formel: HCl
Molecular Weight: 36,46 g/mol
Dichte: 1,08 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Titer 4.9 - 5.1 mol/l
Appearance Passes test
Free bromine or chlorine Passes test
Identification Passes test
Colouration ≤ 10 APHA
Evaporation residue ≤ 100 ppm
Free chlorine ≤ 1 ppm
Heavy metals (as Pb) ≤ 1 ppm
Ignition residue (SO₄) ≤ 80 ppm
SO₃ (Sulphite) ≤ 10 ppm
SO₄ (Sulphate) ≤ 5 ppm
As (Arsenic) ≤ 1.5 ppm
Residual metal catalysts Unlikely to be present
Residual solvents (ICH Q3C) Excl.by manufact. process
Endotoxin ≤ 0.50 EU/ml
Filtered through 0.2 μm Confirmed

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR