N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

85432-10ML 85432-1ML 85432-10X1ML
85432-10MLEA 162 EUR
85432-10ML 85432-1ML 85432-10X1ML
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) silylating mixture II according to Horning, LiChropur™ zur GC-Derivatisierung, Supelco®
N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole (BSA+TMCS+TMSI) mixture
Flammpunkt: 4 °C
UN: 2924
ADR: 3 (8) II

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Faint Yellow to Yellow
Appearance (Form) Liquid
Refractive index N20/D 1,434 - 1,438
Proton NMR Spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR