1,3-Dimethyl-2-imidazolidinon ≥99,5%, PESTINORM® für die Headspace-Gaschromatographie

Lieferant: VWR Chemicals
Gefahr

85448.290EA 1340 EUR
85448.290
1,3-Dimethyl-2-imidazolidinon ≥99,5%, PESTINORM® für die Headspace-Gaschromatographie
1,3-Dimethyl-2-imidazolidinon
Formel: C₅H₁₀N₂O
Siedepunkt: 225,5 °C (1013 hPa)
Schmelzpunkt: 8 °C
Dichte: 1,04 g/cm³ (20 °C)
Flammpunkt: 93 °C
MDL: MFCD00003188
CAS-Nummer: 80-73-9
EINECS: 201-304-8
UN: 2810
ADR: 6.1,III
Merck Index: 13,03275

Bestellen

Spezifikation Testergebnisse

Identity (IR) Complying
Purity (GC on anhydrous basis) min. 99.5%
Water (KF) max. 0.03% w/w
Color (APHA Pt-Co) max. 10
GC-HS (Class 1 solvents) max. 1 µg/g
GC-HS (Class 2 solvents) max. 10 µg/g
GC-HS (Class 3 solvents) max. 50 µg/g
GC-HS (DMF) max. 20 µg/g
GC-HS (Each unknown, as MIBK) max. 10 µg/g
Suitable as solvent for the analysis of residual solvents of ICH classes 1,2 and 3 according to Ph Eur and USP.
Formic acid, acetic acid and cumene are not detected.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR