Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) Dihydrat, Supelco®
Lieferant: Merck
Synonyme:
Spermine Dihydrate
M-Clarity™ Qualitätsstufe = MQ100
85588-5G
85588-5GEA
194
EUR
85588-5G
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) Dihydrat, Supelco®
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) Dihydrat
Formel:
C₁₀H₂₆N₄·2H₂O |
CAS-Nummer:
403982-64-9 EINECS: 200-754-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...