pH-Pufferlösung nach Reag. Ph. Eur. 4000600
Lieferant: VWR Chemicals
85669.290EA 90 EUR
85669.290
pH-Pufferlösung nach Reag. Ph. Eur. 4000600
Puffer pH-Puffer
Contents: ammonium acetate R / water R / hydrochloric acid R1.
  • Ph Eur N 4000600
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.
Bestellinformation: Normalerweise auf Anfrage gefertigt. Bitte wenden Sie sich an den Kundendienst.
Lieferumfang: Delivery with certificate of analysis.