Chromate test strips, Dosatest
Lieferant: VWR Chemicals
85695.601EA 42.2 EUR
85695.601
Chromate test strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of chromate.
  • Ideal for rapid and easy analysis of chromate directly on-site
  • No maintenance or calibration is required
Colour change: White to violet.
Lieferumfang: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box