Nickel Test Strips, Dosatest
Lieferant: VWR Chemicals
85697.601EA
32.9
EUR
85697.601
Nickel Test Strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of nickel.
- Ideal for rapid and easy analysis of nickel directly on-site
- No maintenance or calibration is required
Colour change: White to bright red.
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.
Bitte beachten Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.