VWR®, Sulphite test strips, Dosatest
Lieferant: VWR Chemicals
85699.601EA
32.9
EUR
85699.601
VWR®, Sulphite test strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of sulphite.
- Ideal for rapid and easy analysis of sulphite directly on-site
- No maintenance or calibration is required
Colour change: White to salmon
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.
Bitte beachten Acid solutions such as white wine have to be buffered with solid sodium acetateas the the test paper does not react in acidic media.