VWR®, Zinc Test Strips, Dosatest
Lieferant: VWR Chemicals
85700.601EA 49.4 EUR
85700.601
VWR®, Zinc Test Strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of zinc.
  • Ideal for rapid and easy analysis of zinc directly on-site
  • No maintenance or calibration is required
Colour change: Orange to red
Lieferumfang: Delivered as 100 test strips in a tube with reagents measuring vessel and instruction leaflet.
Bitte beachten Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.