Sulphate test strips, Dosatest, VWR®
Lieferant: VWR Chemicals
85708.601EA 34.6 EUR
85708.601
Sulphate test strips, Dosatest, VWR®
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of sulphate.
  • Ideal for rapid and easy analysis of sulphate directly on-site
  • No maintenance or calibration is required
Colour change: Red to yellow
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.
Bitte beachten If the sample solution has a pH value of 4 to 8, the reaction will take place without interferences. Strongly acid solutions have to be buffered with sodium acetate, and alkaline with tartaric acid.