Arsenic test strips, Dosatest
Lieferant: VWR Chemicals
85710.601EA 88.4 EUR
85710.601
Arsenic test strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of arsenic.
  • Ideal for rapid and easy analysis of arsenic directly on-site
  • No maintenance or calibration is required
Colour change: White to yellow-brown
Lieferumfang: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box
Bitte beachten Each test field on the test strips contains mercury(II) bromide. Toxic in contact with skin