Quaternary ammonium test strips, Dosatest, VWR®
Lieferant: VWR Chemicals
85711.601EA
31.7
EUR
85711.601
Quaternary ammonium test strips, Dosatest, VWR®
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of QUAT.
- Ideal for rapid and easy analysis of QUAT ammonium directly on-site
- No maintenance or calibration is required
Colour change: Yellow to blue-green
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.