Brom-Lösung R Reag. Ph. Eur. 1012401

Lieferant: VWR Chemicals
Gefahr

85715.180EA 110 EUR
85715.180
Brom-Lösung R Reag. Ph. Eur. 1012401
Bromine

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water. Short shelf life of max. 3 months


Starting materials: Bromine R / Potassium bromide R / Water R

Formel: Br₂
Molecular Weight: 159,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010896
CAS-Nummer: 7726-95-6
UN: 1744
ADR: 8(6.1),I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR