Bromwasser (gesättigte Lösung von Brom in Wasser) Reag. Ph. Eur. 1012402

Lieferant: VWR Chemicals
Gefahr

85716.180EA 98.9 EUR
85716.180
Bromwasser (gesättigte Lösung von Brom in Wasser) Reag. Ph. Eur. 1012402
Bromwasser (gesättigte Lösung von Brom in Wasser)

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.

Formel: Br₂
Molecular Weight: 159,81 g/mol
MDL: MFCD00010896
CAS-Nummer: 7726-95-6
UN: 1744
ADR: 8 (6.1) ,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR