Magnesiumnitrat 10 g/L (99,999%), ARISTAR® Matrixmodifizierlösung, für die Graphitofen-AAS
Lieferant: VWR Chemicals
85780.154EA
108
EUR
85780.154
Magnesiumnitrat 10 g/L (99,999%), ARISTAR® Matrixmodifizierlösung, für die Graphitofen-AAS
Magnesium nitrate solutions
The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.
- Guaranteed accuracy: ±1%
- COA incl. up to 70 measured trace impurities in ppm range
VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.
Formel:
Mg(NO₃)₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
13446-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...