1-Butansulfonsäure Natriumsalz ≥99,0% (Trockenmasse), HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
85790.150EA
234
EUR
85790.150
1-Butansulfonsäure Natriumsalz ≥99,0% (Trockenmasse), HiPerSolv CHROMANORM® für die HPLC
1-Butansulfonsäure Natriumsalz
Formel:
C₄H₉NaO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007540 CAS-Nummer: 2386-54-1 |
Spezifikation Testergebnisse
Assay (calculated on dried substance) | ≥ 99,0 % |
Appearance | White powder |
IR Spectrum | Passes test |
Appearance of solution (10 %) | Clear and colourless |
Water | ≤ 1,0 % |
Absorbance (210 nm) (0.5 mol/l) | ≤ 0,1 |
Absorbance (220 nm) (0.5 mol/l) | ≤ 0,06 |
Absorbance (230 nm) (0.5 mol/l) | ≤ 0,04 |
Absorbance (260 nm) (0.5 mol/l) | ≤ 0,02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...