Natrium-1-decansulfonat ≥99,0%, HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
Synonyme:
Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt
85792.150EA
306
EUR
85792.150
Natrium-1-decansulfonat ≥99,0%, HiPerSolv CHROMANORM® für die HPLC
Natrium-1-decansulfonat
Formel:
C₁₀H₂₁NaO₃S Molecular Weight: 244.33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007526 CAS-Nummer: 13419-61-9 |
Spezifikation Testergebnisse
Assay (calculated on dried substance) | ≥ 99,00 % |
Appearance | White to cream powder |
IR Spectrum | Passes test |
Appearance of solution (5 %) | Clear and colourless |
Water | ≤ 1,0 % |
Absorbance (210 nm) (0.2 mol/l) | ≤ 0,05 |
Absorbance (220 nm) (0.2 mol/l) | ≤ 0,03 |
Absorbance (230 nm) (0.2 mol/l) | ≤ 0,02 |
Absorbance (260 nm) (0.2 mol/l) | ≤ 0,02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...