Lithiumdodecylsulfat ≥98,5%, HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
|
Gefahr
|
85793.150EA
306
EUR
85793.150
Lithiumdodecylsulfat ≥98,5%, HiPerSolv CHROMANORM® für die HPLC
Lithiumdodecylsulfat
Formel:
C₁₂H₂₅LiO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007467 CAS-Nummer: 2044-56-6 UN: 1325 ADR: 4.1,III |
Spezifikation Testergebnisse
Assay (C12) | ≥ 98,50 % |
Appearance | White fine crystals |
IR Spectrum | Passes test |
Loss on drying (120°C; under vacuum) | ≤ 0,50 % |
Appearance of solution (1 %; ethanol) | Clear and colourless |
Appearance of solution (3 %) | Clear and colourless |
Cl (Chloride) | ≤ 100,0 ppm |
Absorbance (260 nm) (3 %) | ≤ 0,100 |
Absorbance (280 nm) (3 %) | ≤ 0,075 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...