Kaliumchlorid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Lieferant: VWR Chemicals
Synonyme:
Potassiumchloride
85809.290EA
26.5
EUR
85809.290
Kaliumchlorid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 Schmelzpunkt: 773 |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...