Ethanol eurodenaturiert ≥99%, TechniSolv® (1:1:1)
Lieferant: VWR Chemicals
|
|
Completely denatured alcohol (CDA).
The (1:1:1) denaturant is in compliance with European legislation and comprises methyl ethyl ketone (MEK) 1% v/v and 2- propanol (IPA) 1% v/v and denatonium benzoate 1 g/100 litres. (All figures based on the ethanol content, not on any percentage of water present)
Denatonium benzoate (a tasting agent) is also known by several trade names including, Bitrex, BITTERANT-b, BITTER+PLUS.
Formel:
H₃CCH₂OH |
CAS-Nummer:
64-17-5 UN: 1170 ADR: 3,II |
Spezifikation Testergebnisse
Assay (V/V) | min. 97.2% |
Denatonium benzoate (ppm) | 12.3 - 14.7 ppm |
Methyl ethyl ketone (V/V) | 0.97 - 1.13 % |
2-Propanol (V/V) | 0.97 - 1.13% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...