Eriochromblauschwarz B (C.I. 14640)

Lieferant: VWR Chemicals
Achtung

85900.150EA 29.75 EUR
85900.150
Eriochromblauschwarz B (C.I. 14640)
Eriochromblauschwarz B
Formel: C₂₀H₁₃N₂NaO₅S
Molecular Weight: 416,39 g/mol
MDL: MFCD00003934
CAS-Nummer: 3564-14-5

Bestellen

Spezifikation Testergebnisse

Appearance black powder
Absorption maximum λmax (in water) 520 - 540 nm
Absorbance A (at λmax; c=20 mg/l in water + 10ml HCl 1 mol/l; 1 cm) 0,2 - 0,6
Loss on drying (110 °C) max. 10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR