Ethanol 20% für die biopharmazeutische Produktion

Lieferant: VWR Chemicals
Achtung

85901.290EA 62.8 EUR
85901.290 85901.410 85901.550
Ethanol 20% für die biopharmazeutische Produktion
Ethanol
Formel: C₂H₅OH
Dichte: 0,9686 g/cm³ (20°C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,III
REACH: 01-2119457610-43-XXXX

Bestellen

Spezifikation Testergebnisse

assay 19,5 - 20,5%
appearance passes test
evaporation residue ≤ 10 pmm
residual solvents (ICHQ3C) excluded by manufacturing process
residual metal catalysts unlikely to be present
endotoxin ≤ 2,5 EU/ml
TAMC ≤ 10 CFU/ml
filtered through 0,2µm confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR