Methoxypolyethylene glycol 5,000 acetic acid N-succinimidyl ester ≥80%, Supelco®

Lieferant: Merck

Synonyme: m-PEG-NHS ester, mPEG-Succinimidyl Carboxy Methyl ester) Mn 2000

85973-1G
85973-1GEA 519 EUR
85973-1G
Methoxypolyethylene glycol 5,000 acetic acid N-succinimidyl ester ≥80%, Supelco®
Methoxypolyethylene glycol 5,000 acetic acid N-succinimidyl ester

Verpackung: Glass bottle

Lagertemperatur: Gefrierschrank
MDL: MFCD01867746
CAS-Nummer: 92451-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c875c10a7c5f8077c787b699c8b0b989498dddc&vid=f295017459c0552ea31edc43e839b91089af1659&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR