Dodecyltrimethylammoniumbromid ≥99%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
|
Synonyme:
Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
85982.180EA
194
EUR
85982.180
Dodecyltrimethylammoniumbromid ≥99%, GPR RECTAPUR®
Dodecyltrimethylammonium bromide
Formel:
C₁₅H₃₄BrN Molecular Weight: 308,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011767 CAS-Nummer: 1119-94-4 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Assay | Min. 99 % |
IR Spectrum | Conforms to structure |
Colouration (10 %; water) | Max. 100 APHA |
Solubility in water (10 %) | Clear to slightly hazy |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...