1-Methylimidazol ≥99,0% (durch GC), GPR RECTAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyme: N-Methylimidazol, NMI, 1-Methyl-1H-imidazol, N-Methyl-1H-imidazol

85994.180EA 32.6 EUR
85994.180 85994.290
1-Methylimidazol ≥99,0% (durch GC), GPR RECTAPUR®
1-Methylimidazol
Formel: C₄H₆N₂
MDL: MFCD00005292
CAS-Nummer: 616-47-7
UN: 2922
ADR: 8 (6.1),II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colourless to yellow liquid
Assay (GC) min. 99.0 %
Density (d20°C/ 4°C) 1.034 - 1.036
Water max. 0.5 %
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR