1-Methylimidazol ≥99,0% (durch GC), GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
N-Methylimidazol, NMI, 1-Methyl-1H-imidazol, N-Methyl-1H-imidazol
85994.180EA
32.6
EUR
85994.180
85994.290
1-Methylimidazol ≥99,0% (durch GC), GPR RECTAPUR®
1-Methylimidazol
Formel:
C₄H₆N₂ |
MDL:
MFCD00005292 CAS-Nummer: 616-47-7 UN: 2922 ADR: 8 (6.1),II |
Spezifikation Testergebnisse
Appearance | Clear colourless to yellow liquid |
Assay (GC) | min. 99.0 % |
Density (d20°C/ 4°C) | 1.034 - 1.036 |
Water | max. 0.5 % |
Identity (IR) | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...