Leukocyte Alkaline Phosphatase Kit based on naphthol AS-MX phosphate and fast blue RR salt (with citrate), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ600
Leukocyte Alkaline Phosphatase (LAP) kits are intended for the semi-quantitative demonstration of alkaline phosphatase activity in white blood cells.
- Suitable for haematology
- Suitable for histology
Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-MX phosphate alkaline solution with fast blue RR salt. The resulting blue, insoluble, granular dye deposit indicates sites of alkaline phosphatase activity.
Kit component also available separately
Hematoxylin Solution, Mayer’s 100 mL (MHS1)
Naphthol AS-MX phosphate 20 mL (855)
Citrate Concentrated Solution 20 mL (854C)
Fast Blue RR Salt, Preweighed Capsules 10 capsules (FBS25)
![]() |