Naphthol AS-BI Alkaline solution, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

861-10ML
861-10MLEA 179 EUR
861-10ML
Naphthol AS-BI Alkaline solution, Sigma-Aldrich®
Naphthol AS-BI Alkaline solution

Kit component for Sigma Alkaline Phosphatase Procedure, kits 86C and 86R

Lagertemperatur: Kühlschrank
MDL: MFCD00283933
ADR: 8,III
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR