Kaliumhexafluorozirkonat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 16923-95-8
Molecular Formula: K2ZrF6
Formula Weight: 283.41
Physical Form: -10 Mesh
Density: 3.48
Solubility: Slightly soluble in cold water, soluble in hot water
Merck Reference: 14,7639
UN No.: 3288
MDL No.: MFCD00042547
For manufacture of metallic zirconium
Formel:
F₆K₂Zr Dichte: 3,48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042547 CAS-Nummer: 16923-95-8 EINECS: 240-985-6 UN: 3288 ADR: 6.1,III Merck Index: 14,7639 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...