Azure II, Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: C.I. 52010/52015, 3,7 Bis(dimethylamino)phenothiazin-5-ium chloride and 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium chloride, Azur B/Methylene Blue 1:1, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride mixt. with 3-(dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride(1:1), Methylene Azure II

861065-25G
861065-25GEA 102 EUR
861065-25G
Azure II, Pulver, Sigma-Aldrich®
Azure II

Azure II has been used in the Richardson′s stock solution for the staining of lysosomal glycogen. It has also been used in the periodic acid schiff staining procedure for the staining of glycogen, nuclei and cytoplasm.

Formel: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
Molecular Weight: 625,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00080716
CAS-Nummer: 37247-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5a8f72d21506a9e3b3c05e38d7745f2496716b8&vid=cf8dcca41a1624592f529e66ab7183fcc804e441&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR