Bismarckbraun G, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ100
861111-100G
861111-25G
861111-100GEA
133
EUR
861111-100G
861111-25G
Bismarckbraun G, Sigma-Aldrich®
Bismarckbraun G
Formel:
C₁₈H₁₈N8·2HCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012977 CAS-Nummer: 10114-58-6 EINECS: 233-314-3 Merck Index: 12,01294 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...