Erioglaucine disodium salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

861146-25G 861146-5G
861146-25GEA 130 EUR
861146-25G 861146-5G
Erioglaucine disodium salt, Sigma-Aldrich®
Erioglaucine disodium salt

Erioglaucine disodium salt has been used as a coloring dye to determine the feeding behaviour in Drosophila. It has been used as a coloring dye for determining the number of egg masses of Meloidogyne incognita.


  • Erioglaucine is an acid aminotriphenylmethane dye with blue color

Formel: C₃₇H₃₄Na₂N₂O₉S₃
Lagertemperatur: Room Temperature
MDL: MFCD00012141
CAS-Nummer: 3844-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43c8f710e793969cc7ac8c2c111bf713d29a7e64&vid=d448e3ebbaef02586cee253abb680b7b8e61922f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Purple to Dark Purple
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength
c = 0.005 g/l; Water
627 - 631 nm
Extinction Coefficient ≥10000
Wavelength 306 - 310 nm
Thin Layer Chromatography Consistent with Past Lots
Solubility (Turbidity)
c = 1 mg/ml; Water
Clear
Solubility (Colour) Dark Blue
Carbon 39.2 - 72.9 %
Nitrogen 2.4 - 4.6 %
Dye Content Based on Nitrogen Content


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR