Thymolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. Indikator

Lieferant: VWR Chemicals

Synonyme: 2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid

86157.050EA 37.02 EUR
86157.050 86157.150 86157.180
Thymolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. Indikator
Thymolphthalein
Formel: C₂₈H₃₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00005909
CAS-Nummer: 125-20-2

Bestellen

Spezifikation Testergebnisse

Identity (UV-VIS Spectroscopy) Passes Test
Appearance of Solution Passes Test
pH Range (Colourless to Blue) 8.8 - 10.5
Absorption Maximum λ (pH 10,5) 592 - 596
Absorptivity (A1%/1 cm; λ max , 0.01 g/l, pH 10.5, on Dried Sample) 800 - 900
Loss on Drying (110 °C) ≤1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR