Phenolphthalein 0,375% in Methanol Indikator

Lieferant: VWR Chemicals
Gefahr

Synonyme: 3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon

86196.230EA 47 EUR
86196.230
Phenolphthalein 0,375% in Methanol Indikator
Phenolphthalein
Formel: C₂₀H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00005913
CAS-Nummer: 77-09-8
UN: 1230
ADR: 3 (6.1),II

Bestellen

Spezifikation Testergebnisse

Application test Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR