Dichlor-1,3,5-triazintrion Natriumsalz Dihydrat, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: NaDCC, Troclosennatrium Dihydrat, Dichlorisocyanursäure Natriumsalz Dihydrat

86197.180EA 112 EUR
86197.180
Dichlor-1,3,5-triazintrion Natriumsalz Dihydrat, AnalaR NORMAPUR® analytisches Reagens
Dichloroisocyanuric acid sodium salt dihydrate
Formel: C₃Cl₂N₃NaO₃·2H₂O
Molecular Weight: 255,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149404
CAS-Nummer: 51580-86-0
EINECS: 220-767-7
UN: 3077
ADR: 9,

Bestellen

Spezifikation Testergebnisse

Assay (iodometric) (calculated on dried) ≥ 98.0 %
Heavy metals (as Pb) ≤ 10 ppm
Loss on drying 9.0 - 16.0 %
Fe (Iron) ≤ 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR