Kaliumdisulfat ≥97,5%, AnalaR NORMAPUR® ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: di-Kaliumdisulfat, Kaliumpyrosulfat

86198.230EA 95.6 EUR
86198.230 86198.290 86198.360 86198.460
Kaliumdisulfat ≥97,5%, AnalaR NORMAPUR® ACS analytisches Reagens
Kaliumdisulfat
Formel: K₂O₇S₂
Schmelzpunkt: 335 °C
Dichte: 2,280 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011385
CAS-Nummer: 7790-62-7
EINECS: 232-216-8
UN: 2923
ADR: 8 (6.1),I
Merck Index: 13,07749

Bestellen

Spezifikation Testergebnisse

Assay (acidimetric) 97.5 - 100.0%
Appearance White granules
Acidity (as H2SO4) 37.5 - 38.6%
Heavy metals (as Pb) max. 10 ppm
Insoluble and ny NH4OH precipitated matter max. 100 ppm
Total N (Nitrogen) max. 20 ppm
Water max. 2.5%
Cl (Chloride) max. 5 ppm
PO4 (Phosphate) max. 10 ppm
Al (Aluminium) max. 10 ppm
As (Arsenic) max. 5 ppm
Ca (Calcium) max. 20 ppm
Fe (Iron) max. 5 ppm
Mg (Magnesium) max. 5 ppm
Na (Sodium) max. 100 ppm
Conform to ACS passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR