FRV-Alkaline solution, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
862-10ML
862-10MLEA
190
EUR
862-10ML
FRV-Alkaline solution, Sigma-Aldrich®
FRV-Alkaline solution
For use with Sigma kit 86R, for the demonstration of alkaline phosphatase in granulocytes. The procedure is often referred to as the LAP procedure.
Fast red violet LB base, 5 mg/mL, in hydrochloric acid, 0.4 mol/L, with stabilizer.
Lagertemperatur:
Kühlschrank |
ADR:
8 III |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...