Test mixture 2 for apolar capillary columns according to Grob, for GC
Lieferant: Merck
Mixture of D(−)-2,3-butanediol, decane, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylcaproic acid, methyl decanoate, methyl laurate, methyl undecanoate, 1-octanol and dodecane in hexane and methylene chloride
Storage temperature: 2 to 8 °C