Dibenzyl chloromethyl phosphate ≥97,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®

Lieferant: Merck
Achtung

86546-1G
86546-1GEA 565 EUR
86546-1G
Dibenzyl chloromethyl phosphate ≥97,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
Dibenzyl chloromethyl phosphate

used in the synthesis of water-soluble prodrugs of lipophilic alcohols, phenols and amines. This compound is an improved derivative of the well known Di-tert-butyl chloromethyl phosphate. So it can be detected more easily by HPLC-UV, because of higher stability, higher yields and better UV-activity.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₅H₁₆ClO₄P
Lagertemperatur: Gefrierschrank
MDL: MFCD16652586
CAS-Nummer: 258516-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf157ec43b9dccfeb2317faa7d1efd42209ab7d1&vid=8f49abf2e639d3f5c803387c3f571e9e7f3163ca&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless to Light Yellow
Appearance (Form) Liquid or Viscous Liquid
Purity (HPLC Area %) ≥ 97.0 %
Proton NMR spectrum Conforms to structure
Phosphorus 31 NMR spectrum Conforms to structure
Expiry period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR