Tetraethylammoniumhexafluorophosphat, Supelco®

Lieferant: Merck

Synonyme: Tetraethylammoniumhexafluorophosphate

M-Clarity™ Qualitätsstufe = MQ100

86625-25G
86625-25GEA 683 EUR
86625-25G
Tetraethylammoniumhexafluorophosphat, Supelco®
Tetraethylammonium hexafluorophosphate
Formel: C₈H₂₀F₆NP
Molecular Weight: 275,22 g/mol
MDL: MFCD00043179
CAS-Nummer: 429-07-2
EINECS: 207-056-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02c273f048211278c8910c2707201e5105fe2adf&vid=a9239e40199f9d00519e0d550b2868d305eb07d0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR