Xylol (Isomerengemisch) ≥98,5% (durch GC), AR® ACS für die Histologie, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

8668-26EA 0 EUR
8668-26 8668-22 8668-06
Xylol (Isomerengemisch) ≥98,5% (durch GC), AR® ACS für die Histologie, Macron Fine Chemicals™
Xylol (Isomerengemisch)

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formel: C₆H₄(CH₃)₂
Siedepunkt: 140 °C (1013 hPa)
Schmelzpunkt: –34 °C
Dichte: 0,86…0.88 g/cm³ (20 °C)
Flammpunkt: 29 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (m,p,o-xylenes and ethylbenzene) (by GC) ≥98.5 %
ACS - Color (APHA) ≤10
ACS - Ethylbenzene (C₆H₅C₂H₅) ≤25 %
ACS - Particulate Contamination Passes Test
ACS - Residue after Evaporation ≤0.002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Sulfur Compounds (as S) ≤0.0005 %
ACS - Water (H₂O) ≤0.02 %
Appearance (clear, colorless liquid)
Date Received: / /
Date Opened: / /


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR