Tetrabutylammonium methanesulphonate ≥97,0% (durch titrimetrische Analyse), Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
86877-10G
86877-10GEA
95
EUR
86877-10G
Tetrabutylammonium methanesulphonate ≥97,0% (durch titrimetrische Analyse), Supelco®
Tetrabutylammonium methanesulphonate
Formel:
[CH₃CH₂CH₂CH₂]₄N(CH₃SO₃) Molecular Weight: 337,56 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00043226 CAS-Nummer: 65411-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...